missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human ICAM-3/CD50 Fc Chimera Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Konjugat Unconjugated
Zusammensetzung Lyophilized from a 0.2μm filtered solution in PBS.
Gen-ID (Entrez) 3385
Molekulargewicht 76.7 kDa (monomer)
Name ICAM-3/CD50 Fc Chimera
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Human ICAM-3/CD50 Fc Chimera Protein is derived from NS0. The Recombinant Human ICAM-3/CD50 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

Unconjugated
3385
ICAM-3/CD50 Fc Chimera
Mouse myeloma cell line,NS0-derived human ICAM-3/CD50 protein, (N-terminus) Human ICAM-3 (Gln30-His485) Accession &Num; CAA49473 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Lyophilized from a 0.2μm filtered solution in PBS.
76.7 kDa (monomer)
Yes
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen