Learn More
R&D Systems™ Human IFN-gamma ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
Marke:  R&D Systems™ XEL285
Weitere Details : Netto-Gewicht : 0.01000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13413296
  • 3034.08 € / 6 Stück
Voraussichtlicher Liefertermin 22-10-2019
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beinhaltet:
  • Microplate
  • Biotinylated Detection Antibody
  • Streptavidin conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • Positive Control
Beschreibung
Beschreibung
Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells
Spezifikation
Spezifikation
IFN-gamma
Quantitative determination of the frequency of cells releasing human IFN-g
Human IFN-gamma ELISpot Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Human
Human IFN-gamma Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Human IFN-gamma Positive Control
Whole Cells
6 x 96-well plates