Learn More
R&D Systems™ Human IFN-gamma ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
N.V.
Spezifikation
Antigen IFN-gamma
Kreuzreaktivität Human
Produkttyp Human IFN-gamma ELISpot Kit
Enthält Human IFN-gamma Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Human IFN-gamma Positive Control
Probentyp Whole Cells
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously. Complete ELISpot kits are ready-to-r
Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • Hig
Spezifikation
Spezifikation
IFN-gamma
Human IFN-gamma ELISpot Kit
Whole Cells
Human
Human IFN-gamma Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Human IFN-gamma Positive Control
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Produktzertifizierungen