missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-2 (Cys145Ser) Recombinant Protein

Marke:  R&D Systems™ 1081-IL-020

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13418523

  • 219.12 € / 20 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Ala21-Thr153 (Cys145Ser), with and without an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.5 kDa
Formulation: Supplied as a 0.2μm filtered solution in Sodium Acetate.
Entrez Gene ID: 3558
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Sodium Acetate with BSA as a carrier protein.
15.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3558
IL-2 (Cys145Ser)
20μg
Unlabeled
Videos