missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-34 Recombinant Protein

Marke:  R&D Systems™ 5265-IL-010/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415374

  • 432.92 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Asn21-Pro242, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 26.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Entrez Gene ID: 146433
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
26.1 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
146433
IL-34
10μg
Unlabeled
Videos