Learn More
R&D Systems™ Human IL-36 alpha/IL-1F6 (aa 6-158) Recombinant Protein
Marke:  R&D Systems™ 6995-IL-010/CF
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13416024
  • 444.62 € / 10 Mikrogramm
Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Measured by its ability to induce IL-8 secretion in A431 human epithelial carcinoma cells. The ED 50 for this effect is 4-24 ng/mL.
Spezifikation
Spezifikation
Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween® 20 with Trehalose.
17 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Monomer
Yes
27179
IL-36 alpha/IL-1F6 (aa 6-158)
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Reconstitute at 100μg/mL in Water.
Videos