missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-36 gamma/IL-1F9 (aa 18-169) Recombinant Protein

For use in cell or tissue culture applications.

Marke:  R&D Systems™ 6835-IL-010/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415964

  • 432.92 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 22-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Source: E. coli-derived, Ser18-Asp169
  • Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
  • Molecular Weight: 17 kDa
  • Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, TCEP and EDTA.
  • Entrez Gene ID: 56300
  • Regulatory Status: RUO
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, TCEP and EDTA.
17 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
56300
IL-36 gamma/IL-1F9 (aa 18-169)
10μg
Unlabeled
Videos