missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-4 R alpha Recombinant Protein

Marke:  R&D Systems™ 230-4R-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13410994

  • 401.72 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 24-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gly24-His232
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3566
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
24 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
3566
IL-4 R alpha
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos