Learn More
R&D Systems™ Human KGF/FGF-7 Recombinant Protein
Marke:  R&D Systems™ 251-KG-010
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411384
  • 410.82 € / 10 Mikrogramm
Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Measured in a cell proliferation assay using 4MBr-5 rhesus monkey epithelial cells. Rubin, J.S. et al. (1989) Proc. Natl. Acad. Sci. USA 86:802. The ED 50 for this effect is typically 6-60 ng/mL. The specific activity of recombinant human KGF/FGF-7 is approximately 1.3 x 10 3 U/µg, which is calibrated against recombinant human KGF/FGF-7 WHO Standard (NIBSC code: 03/150).
Spezifikation
Spezifikation
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA with BSA as a carrier protein.
19 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
2252
KGF/FGF-7
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.