missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Lefty-A Recombinant Protein

Marke:  R&D Systems™ 746-LF-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416204

  • 275.89 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ability to induce cell death using Mv1Lu mink lung epithelial cells. The ED 50 for this effect is 0.5-3 µg/mL.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
39 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line, NS0-derived Mouse Lefty-1 Propeptide (Leu22-Arg77) Accession # Q64280 HHHHHH Human Lefty-A (Phe78-366) Accession # O00292
Unlabeled
Reconstitute at 0.5mg/mL in sterile PBS.
7044
Lefty-A
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos