missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human LIGHT/TNFSF14 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Marke:  R&D Systems™ 664-LI-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415894

  • 487.52 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 01-08-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The Recombinant Human LIGHT/TNFSF14 Protein is derived from NS0. The Recombinant Human LIGHT/TNFSF14 Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
20.9 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line, NS0-derived Met 10-His tag GGGSGGGSGGGSIEGR Human Light (Asp74-Val240) Accession # O43557
Unlabeled
8740
LIGHT/TNFSF14
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos