missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human MD-1 Recombinant Protein

Marke:  R&D Systems™ 925-MD-050/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416424

  • 333.97 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Gly21-Ser162
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 32.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 9450
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
32.5 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
9450
MD-1
50μg
Unlabeled
Videos