missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Mouse/Rat/Canine/Equine BDNF Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Marke:  R&D Systems™ 248-BD-005

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13411324

  • 386.61 € / 5 Mikrogramm

Voraussichtlicher Liefertermin 22-10-2019

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The Recombinant Human BDNF Protein is derived from Sf 21 (baculovirus). The Recombinant Human BDNF Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Sodium Citrate and NaCl with BSA as a carrier protein.
13.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
627
BDNF
5μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Dokumentation