missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human MSP/MST1 (Cys672Ala) Recombinant Protein

Marke:  R&D Systems™ 4306-MS-010

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414534

  • 421.22 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Chinese Hamster Ovary cell line, CHO-derived, Gln19-Arg483 (alpha) & Val484-Gly711 (Cys672Ala) (beta)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 53 kDa ( alpha-chain), 25 kDa (beta-chain)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 4485
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
53 kDa ( alpha-chain), 25 kDa (beta-chain)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
4485
MSP/MST1 (Cys672Ala)
10μg
Disulfide-linked heterodimer
Yes
Videos