Learn More
R&D Systems™ Human Neuregulin-1 alpha/NRG1 alpha EGF Domain Recombinant Protein
Marke:  R&D Systems™ 296-HR-050
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13412164
  • 275.89 € / 50 Mikrogramm
Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Measured in a serum-free cell proliferation assay using MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED 50 for this effect is 10-50 ng/mL.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
3084
Neuregulin-1 alpha/NRG1 alpha EGF Domain
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos