missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Neuregulin-1 beta 1/NRG1 beta 1 Extracellular Domain Recombinant Protein

Marke:  R&D Systems™ 377-HB-050/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413594

  • 487.52 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Ser2-Lys246
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 26.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 3084
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
26.9 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3084
Neuregulin-1 beta 1/NRG1 beta 1 Extracellular Domain
50μg
Unlabeled
Videos