missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human NGF R/TNFRSF16 Fc Chimera Recombinant Protein

Marke:  R&D Systems™ 367-NR-050/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413414

  • 295.69 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ability to inhibit beta-NGF-dependent proliferation of TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED 50 for this effect is 0.2-0.6 µg/mL.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
51 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Sf 21 (baculovirus)
Disulfide-linked homodimer
Yes
4804
NGF R/TNFRSF16 Fc Chimera
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos