missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Pref-1/DLK-1/FA1 Recombinant Protein

Marke:  R&D Systems™ 1144-PR-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13418623

  • 353.77 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Ala24-Pro297 (Arg248Pro) (Lys295Ser) with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 29.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 8788
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
29.7 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
8788
Pref-1/DLK-1/FA1
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos