Learn More
R&D Systems™ Human TGF-beta 1.2 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke:  R&D Systems™ 304-B3-001/CF
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13412314
  • 520.38 € / 1 Mikrogramm
Voraussichtlicher Liefertermin 22-10-2019
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
The Recombinant Human TGF-beta 1.2 Protein is derived from Sf 21 (baculovirus). The Recombinant Human TGF-beta 1.2 Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
TGF-beta 1.2
1μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
12.8 kDa (TGF-beta1) & 12.7 kDa (TGF-beta2)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked heterodimer
Yes