Learn More
R&D Systems™ Human TGF-beta 1.2 Recombinant Protein
Marke:  R&D Systems™ 304-B3-001/CF
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13412314
  • 460.22 € / 1 Mikrogramm
Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. Tsang, M. et al. (1995) Cytokine 7:389. The ED 50 for this effect is 0.04-0.08 ng/mL.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
TGF-beta 1.2
1μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
12.8 kDa (TGF-beta1) & 12.7 kDa (TGF-beta2)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked heterodimer