missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TGF-beta 1 Protein

Marke:  R&D Systems™ 100-B-010/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13418293

  • 2441.18 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Human platelet-derived
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 7040
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
TGF-beta 1
10μg
Disulfide-linked homodimer
7040
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
Videos