missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TRAIL R1/TNFRSF10A Fc Chimera Recombinant Protein

Marke:  R&D Systems™ 347-DR-100/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413064

  • 291.73 € / 100 Mikrogramm


Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ability to inhibit TRAIL-mediated cytotoxicity using L-929 mouse fibroblast cells treated with TRAIL. The ED 50 for this effect is 1-3 ng/mL in the presence of 6 ng/mL of Recombinant Human TRAIL/TNFSF10 (Catalog # 375-TEC).
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
43 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Sf 21 (baculovirus)
Disulfide-linked homodimer
Yes
8797
TRAIL R1/TNFRSF10A Fc Chimera
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos