missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TRAIL R3/TNFRSF10C Fc Chimera Recombinant Protein

Marke:  R&D Systems™ 630-TR-100/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415764

  • 353.77 € / 100 Mikrogramm


Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ability to inhibit TRAIL-mediated cytotoxicity using L-929 mouse fibroblast cells treated with TRAIL. The ED 50 for this effect is 15-60 ng/mL in the presence of 12 ng/mL of rhTRAIL (Catalog # 375-TEC).
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
48 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
NS0
Disulfide-linked homodimer
Yes
8794
TRAIL R3/TNFRSF10C Fc Chimera
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos