missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TRAIL R3/TNFRSF10C Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
N.V.
Spezifikation
Gen-ID (Entrez) | 8794 |
---|---|
Molekulargewicht | 48 kDa (monomer) |
Name | TRAILR3/TNFRSF10C |
Menge | 100μg |
Quelle | Mouse myeloma cell line,NS0-derived human TRAIL R3/TNFRSF10C protein, (N-terminus) TRAIL R3 (Ala26-Ala221) Accession &Num; Q05D71 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
Beschreibung
The Recombinant Human TRAIL R3/TNFRSF10C Fc Chimera Protein is derived from NS0. The Recombinant Human TRAIL R3/TNFRSF10C Fc Chimera Protein has been validated for the following applications: Bioactivity.Spezifikation
8794 | |
TRAILR3/TNFRSF10C | |
Mouse myeloma cell line,NS0-derived human TRAIL R3/TNFRSF10C protein, (N-terminus) TRAIL R3 (Ala26-Ala221) Accession &Num; Q05D71 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
48 kDa (monomer) | |
100μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |