missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TRANCE/TNFSF11/RANK L Recombinant Protein, (E. coli-expressed)

Marke:  R&D Systems™ 6449-TEC-010/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415814

  • 432.92 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Arg85-Asp245, with an N-terminal Met
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MES and NaCl.
Entrez Gene ID: 8600
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in MES and NaCl.
18.3 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
8600
TRANCE/TNFSF11/RANK L
10μg
Unlabeled
Videos