missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VEGF 162 Recombinant Protein

Marke:  R&D Systems™ 2347-VE-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13411084

  • 353.77 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. The ED 50 for this effect is 1.0-5.0 ng/mL.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
18.8 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked homodimer
Yes
7422
VEGF 162
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos