Learn More
R&D Systems™ Human VEGF R1/Flt-1 Fc Chimera Recombinant Protein
Marke:  R&D Systems™ 321-FL-050
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13412594
  • 543.42 € / 50 Mikrogramm
Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Measured by its ability to inhibit the VEGF-dependent proliferation of HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED 50 for this effect is 5-30 ng/mL.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl and CHAPS with BSA as a carrier protein.
100 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Sf 21 (baculovirus)
Disulfide-linked homodimer
Unlabeled
2321
VEGF R1/Flt-1 Fc Chimera
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution.
<0.1 EU per 1μg of the protein by the LAL method.