Learn More
R&D Systems™ Human VEGF R1/Flt-1 Fc Chimera Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke:  R&D Systems™ 321-FL-050
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13412594
  • 614.46 € / 50 Mikrogramm
Voraussichtlicher Liefertermin 29-10-2019
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
The Recombinant Human VEGFR1/Flt-1 Fc Chimera Protein is derived from Sf 21 (baculovirus). The Recombinant Human VEGFR1/Flt-1 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl and CHAPS with BSA as a carrier protein.
100 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Sf 21 (baculovirus)
Disulfide-linked homodimer
Unlabeled
2321
VEGF R1/Flt-1 Fc Chimera
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution.
<0.1 EU per 1μg of the protein by the LAL method.
Yes