missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse BMPR-IB/ALK-6 Fc Chimera (NS0) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
N.V.
Spezifikation
Gen-ID (Entrez) | 12167 |
---|---|
Molekulargewicht | 40.6 kDa (monomer) |
Name | BMPR-IB/ALK-6 Fc Chimera |
Menge | 100μg |
Quelle | Mouse myeloma cell line,NS0-derived mouse BMPR-IB/ALK-6 protein, (N-terminus) Mouse BMPR-IB (Leu2-Lys126) Accession &Num; P36898 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
Beschreibung
The Recombinant Mouse BMPR-IB/ALK-6 Fc Chimera (NS0) Protein is derived from NS0. The Recombinant Mouse BMPR-IB/ALK-6 Fc Chimera (NS0) Protein has been validated for the following applications: Bioactivity.Spezifikation
12167 | |
BMPR-IB/ALK-6 Fc Chimera | |
Mouse myeloma cell line,NS0-derived mouse BMPR-IB/ALK-6 protein, (N-terminus) Mouse BMPR-IB (Leu2-Lys126) Accession &Num; P36898 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
40.6 kDa (monomer) | |
100μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |