missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse BMPR-IB/ALK-6 Fc Chimera Recombinant Protein

Marke:  R&D Systems™ 444-BNS-100

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414784

  • 405.62 € / 100 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Leu2-Lys126
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 40.6 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 12167
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
40.6 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
12167
BMPR-IB/ALK-6 Fc Chimera
100μg
Disulfide-linked homodimer
Yes
Videos