missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Cardiotrophin-1/CT-1 Recombinant Protein

Marke:  R&D Systems™ 438-CT-010

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414674

  • 227.05 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 22-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Ser2-Ala203
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 13019
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
21 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
13019
Cardiotrophin-1/CT-1
10μg
Unlabeled
Videos