missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL11/Eotaxin Recombinant Protein

Marke:  R&D Systems™ 420-ME-020/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414364

  • 364.33 € / 20 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, His24-Pro97
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 20292
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
8.4 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
20292
CCL11/Eotaxin
20μg
Unlabeled
Videos