missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL24/Eotaxin-2/MPIF-2 aa 24-119 Recombinant Protein

Marke:  R&D Systems™ 528-MP-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415384

  • 195.36 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Ile24-Val119
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 10.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 56221
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
10.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
56221
CCL24/Eotaxin-2/MPIF-2 aa 24-119
25μg
Unlabeled
Videos