missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL28 Recombinant Protein

Marke:  R&D Systems™ 533-VI-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415404

  • 277.21 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Ile23-Arg130
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
Entrez Gene ID: 56838
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
12.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
56838
CCL28
25μg
Unlabeled
Videos