missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CCL28 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spécification

Gen-ID (Entrez) 56838
Molekulargewicht 12.3 kDa
Name CCL28
Menge 25μg
Quelle E. coli-derived mouse CCL28 protein Ile23-Arg130
Consulter d'autres spécifications

Description

Description

The Recombinant Mouse CCL28 Protein is derived from E. coli. The Recombinant Mouse CCL28 Protein has been validated for the following applications: Bioactivity.
Spécification

Spécification

56838
CCL28
E. coli-derived mouse CCL28 protein Ile23-Arg130
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
12.3 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Vidéos
FDS
Documents

Documents

Product Certifications