missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL9/10/MIP-1 gamma Recombinant Protein

Marke:  R&D Systems™ 463-MG-025

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415014

  • 250.81 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Gln22-Gln122
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 11.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 20308
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
11.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
20308
CCL9/10/MIP-1 gamma
25μg
Unlabeled
Videos