missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CD27 Ligand/TNFSF7 Recombinant Protein

Marke:  R&D Systems™ 783-CL-050

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416314

  • 344.53 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Gln47-Pro195, with an N-terminal Met and 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 21948
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
19.2 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
21948
CD27 Ligand/TNFSF7
50μg
Unlabeled
Videos