missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CXCL16 Chemokine Domain Recombinant Protein

Marke:  R&D Systems™ 503-CX-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415264

  • 364.33 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Asn27-Pro114
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 66102
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
9.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
66102
CXCL16 Chemokine Domain
25μg
Unlabeled
Videos