Learn More
R&D Systems™ Mouse CXCL7/Thymus Chemokine-1 aa 40-113 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marke:  R&D Systems™ 793-CK-025
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13416324
  • 333.97 € / 25 Mikrogramm
Voraussichtlicher Liefertermin 31-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
The Recombinant Mouse CXCL7/Thymus Chemokine-1 (aa 40-113) is derived from E. coli. The Recombinant Mouse CXCL7/Thymus Chemokine-1 (aa 40-113) has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
8.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
57349
CXCL7/Thymus Chemokine-1 aa 40-113
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos