missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CXCL9/MIG Recombinant Protein

Marke:  R&D Systems™ 492-MM-050/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415194

  • 331.33 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Thr22-Thr126
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 17329
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
12 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
17329
CXCL9/MIG
50μg
Unlabeled
Videos