missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse FGF-17 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Gen-ID (Entrez) 14171
Molekulargewicht 22.7 kDa
Name FGF-17
Menge 25μg
Quelle E. coli-derived mouse FGF-17 protein Thr23-Thr216,with an N-terminal Met
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse FGF-17 Protein is derived from E. coli. The Recombinant Mouse FGF-17 Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

14171
FGF-17
E. coli-derived mouse FGF-17 protein Thr23-Thr216,with an N-terminal Met
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
22.7 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen