missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse FGF-6 Recombinant Protein

Marke:  R&D Systems™ 5750-F6-025

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415534

  • 326.05 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Pro39-Ile208
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Ammonium Sulfate, EDTA, DTT.
Entrez Gene ID: 14177
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in MOPS, Ammonium Sulfate, EDTA, DTT with BSA as a carrier protein.
18.8 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
14177
FGF-6
25μg
Unlabeled
Videos