missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse FGF basic Recombinant Protein

Marke:  R&D Systems™ 3139-FB-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412514

  • 360.37 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Rizzino, A. et al. (1988) Cancer Res. 48:4266; Thomas, K. et al. (1987) Methods Enzymol. 147:120. The ED 50 for this effect is 0.3-1.8 ng/mL.
Spezifikation

Spezifikation

Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT.
16.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
14173
FGF basic
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos