missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Follistatin 288 (FS-288) Recombinant Protein

Marke:  R&D Systems™ 769-FS-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416294

  • 481.02 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gly30-Asn317, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 31 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 14313
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
31 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
14313
Follistatin 288 (FS-288)
25μg
Unlabeled
Videos