missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Follistatin 288 (FS-288) Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Gen-ID (Entrez) 14313
Molekulargewicht 31 kDa
Name Follistatin 288 (FS-288)
Menge 25μg
Quelle Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Follistatin protein Gly30-Asn317,with an N-terminal Met
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse Follistatin 288 (FS-288) Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse Follistatin 288 (FS-288) Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

14313
Follistatin 288 (FS-288)
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Follistatin protein Gly30-Asn317,with an N-terminal Met
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
31 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen