missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse GM-CSF Recombinant Protein

Marke:  R&D Systems™ 415-ML-050

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414264

  • 1090.50 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen

Beschrijving

Beschrijving

Measured in a cell proliferation assay using DA3 mouse myeloma cells. Ihle, J.N. et al. (1984) Advances in Viral Oncology. In G. Klein (eds): Raven Press, New York, NY. 4:95. The ED 50 for this effect is 5-30 pg/mL.
Specificatie

Specificatie

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
14.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
12981
GM-CSF
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Video's