missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse GM-CSF Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Gen-ID (Entrez) 12981
Molekulargewicht 14.3 kDa
Name GM-CSF
Quelle E. coli-derived mouse GM-CSF protein Ala18-Lys141,with an N-terminal Met
Lagerungsbedingungen Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse GM-CSF Protein is derived from E. coli. The Recombinant Mouse GM-CSF Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

12981
GM-CSF
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
14.3 kDa
E. coli-derived mouse GM-CSF protein Ala18-Lys141,with an N-terminal Met
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Recombinant
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen