Learn More
R&D Systems™ Mouse IFN-gamma/IL-17 Dual-Color ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
Marke:  R&D Systems™ ELD5007
Weitere Details : Netto-Gewicht : 0.01000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13415145
  • 573.32 € / 1 Set
Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen
Spezifikation
Spezifikation
Diagnostics
Mouse IFNG/IL-17 Dual-Color ELISpot Kit
One 96-well plate
One 96-well plate coated w/mAb for mouse IFNG and mAb for mouse IL-17, Biotin- and HRP-conjugated detection Abs, Streptavidin-AP, Buffers, BCIP/NBT, AEC, Positive controls
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos