missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IGF-I R Recombinant Protein

Marke:  R&D Systems™ 6630-GR-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415884

  • 403.02 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Chinese Hamster Ovary cell line, CHO-derived, Met1-His936, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 104.5 kDa (single chain), 80.7 kDa ( alpha subunit) & 23.2 kDa (beta subunit)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
IGF-I R
25μg
Covalently-linked heterotetramers (2 alpha and 2 beta subunits) & covalently-linked single chain homodimers.
Yes
104.5 kDa (single chain), 80.7 kDa ( alpha subunit) & 23.2 kDa (beta subunit)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
Videos