missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-1 Rrp2/IL-1 R6 Recombinant Protein

Marke:  R&D Systems™ 2354-RP-100/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13411094

  • 403.02 € / 100 Mikrogramm


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Asp22-Arg338 (Ala158Val), with a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 107527
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
36.8 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
107527
IL-1 Rrp2/IL-1 R6
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos