Learn More
R&D Systems™ Mouse IL-17 ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
Marke:  R&D Systems™ XEL421
Weitere Details : Netto-Gewicht : 0.01000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13413306
  • 2579.93 € / 6 Stück
Voraussichtlicher Liefertermin 25-04-2019
Zum Warenkorb hinzufügen
Beinhaltet:
  • Microplate
  • Biotinylated Detection Antibody
  • Streptavidin conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • Positive Control
Beschreibung
Beschreibung
Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells
Spezifikation
Spezifikation
IL-17
Detection of cytokine secreting cells, quantitative sandwich ELISA
Mouse IL-17 ELISpot Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Mouse
Mouse IL-17/IL-17A Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Muse IL-17/IL-17A Positive Control
Whole cells
6 x 96-well plates
Videos