missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-17/IL-17A Recombinant Protein

Marke:  R&D Systems™ 421-ML-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414384

  • 360.37 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 01-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ability to induce IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells. Yao, Z. et al. (1995) Immunity 3:811. The ED 50 for this effect is 0.25-1.25 ng/mL.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
15.5 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Disulfide-linked homodimer
Yes
16171
IL-17/IL-17A
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos